L(+)-Tartaric acid ≥99.0% (by acidimetry) for resolution of racemates, for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.18531.0100
8.18531.0500
8.18531.2500
8.18531.0100
8.18531.0500
8.18531.2500
EM8.18531.0100CS
4536.18
CAD
EM8.18531.0100
EM8.18531.0500
EM8.18531.2500
CA8.18531.0100
CA8.18531.0500
CA8.18531.2500
L(+)-Tartaric acid ≥99.0% (by acidimetry) for resolution of racemates, for synthesis, Sigma-Aldrich®
L(+)-Tartaric acid
Formula:
C₄H₆O₆ Melting Pt: 168…170 °C |
MDL Number:
MFCD00064207 CAS Number: 87-69-4 EINECS: 201-766-0 REACH: 01-2119537204-47 Merck Index: 12,09237 |
|
Specification Test Results
Assay (acidimetric) | ≥ 99.0 % |
Identity (IR) | passes test |
Spec. rotation α²0/D (c=20 in water) | +12.0 - +13.0 ° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...