Magnesium, powder < 0.1 mm for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.18506.0100
8.18506.0500
8.18506.0100
8.18506.0500
EM8.18506.0100CS
7618.76
CAD
EM8.18506.0100
EM8.18506.0500
CA8.18506.0100
CA8.18506.0500
Magnesium, powder < 0.1 mm for synthesis, Sigma-Aldrich®
Magnesium
Formula:
Mg MW: 24.3 g/mol Boiling Pt: 1107 Melting Pt: 651 |
MDL Number:
MFCD00085308 CAS Number: 7439-95-4 UN: 1418 ADR: 4.3 (4.2) II Merck Index: 13,05675 |
Specification Test Results
Solubility | (20 °C) (slow decomposition) |
Melting Point | 651 °C |
Molar Mass | 24.31 g/mol |
Boiling Point | 1107 °C (1013 hPa) |
Vapor Pressure | 0.00013 hPa (325 °C) |
Density | 1.74 g/cm3 (20 °C) |
Ignition Point |
|
Hazard Class | 4.3 |
Hazard Class Description | Dangerous When Wet Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...