(R)-(+)-Limonene for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
(R)-(+)-Dipentene, (R)-(+)-1-Methyl-4-(1-methylvinyl)cyclohexene, (R)-(+)-4-Isopropenyl-1-methyl-1-cyclohexene, D(+)-Limonene, (R)-(+)-p-Mentha-1,8-diene, D-Limonene
M-Clarity™ quality level = MQ200
8.18407.0100
8.18407.0500
8.18407.0100
8.18407.0500
EM8.18407.0100CS
936.94
CAD
EM8.18407.0100
EM8.18407.0500
CA8.18407.0100
CA8.18407.0500
(R)-(+)-Limonene for synthesis, Sigma-Aldrich®
(R)-(+)-Limonene
Formula:
C₁₀H₁₆ MW: 136.24 g/mol |
MDL Number:
MFCD00062991 CAS Number: 5989-27-5 UN: 2052 ADR: 3,III |
Specification Test Results
Solubility | 0.02 g/l (25 °C) |
Melting Point | -74 °C |
Molar Mass | 136.24 g/mol |
Boiling Point | 177 - 178 °C (1013 hPa) |
Vapor Pressure | 2.1 hPa (20 °C) |
Flash Point | 43 °C DIN 51758 |
Refractive Index | 1.4722 (20 °C, 589 nm) |
Explosion Limit | 0.7 - 6.1 %(V) |
Density | 0.84 g/cm3 (20 °C) |
Ignition Point | 255 °C |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...