(1S)-(-)-α-Pinene for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
(-)-α-Pinene, (1S)-(-)-2-Pinene, (-)-Pin-2(3)-ene, L(-)-α-Pinene
M-Clarity™ quality level = MQ200
8.18405.0100
8.18405.0500
8.18405.0100
8.18405.0500
EM8.18405.0100CS
804.22
CAD
EM8.18405.0100
EM8.18405.0500
CA8.18405.0100
CA8.18405.0500
(1S)-(-)-α-Pinene for synthesis, Sigma-Aldrich®
(1S)-(-)-α-Pinene
Formula:
C₁₀H₁₆ MW: 136.24 g/mol |
MDL Number:
MFCD00064145 CAS Number: 7785-26-4 UN: 2368 ADR: 3,III |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | -62 °C |
Molar Mass | 136.23 g/mol |
Boiling Point | 155 - 156 °C |
Vapor Pressure | 5 hPa (25 °C) |
Flash Point | 32 °C |
Refractive Index | 1.446 (20 °C, 589 nm) |
Explosion Limit | 0.8 %(V) |
Density | 0.86 g/cm3 (20 °C) |
Ignition Point | 255 °C |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...