Oxalic acid dihydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Ethanedioic acid dihydrate

M-Clarity™ quality level = MQ200

8.18242.0100 8.18242.1000 8.18242.0100 8.18242.1000
EM8.18242.0100CS 11183.24 CAD
EM8.18242.0100 EM8.18242.1000 CA8.18242.0100 CA8.18242.1000
Oxalic acid dihydrate for synthesis, Sigma-Aldrich®
Oxalic acid dihydrate
Formula: HO₂CCO₂H·2H₂O
MW: 126.07 g/mol
Melting Pt: 101
MDL Number: MFCD00149102
CAS Number: 6153-56-6
UN: 3261
Merck Index: 13,06980
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (manganometric) ≥ 99.0 %
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR