Tin(II) chloride, anhydrous for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Stannous chloride, Tin dichloride

M-Clarity™ quality level = MQ200

8.18150.0100 8.18150.0500 8.18150.0100 8.18150.0500
EM8.18150.0100CS 2558.02 CAD
EM8.18150.0100 EM8.18150.0500 CA8.18150.0100 CA8.18150.0500
Tin(II) chloride, anhydrous for synthesis, Sigma-Aldrich®
Tin (II) chloride
Formula: SnCl₂
MW: 189.62 g/mol
Boiling Pt: 623
Melting Pt: 246
MDL Number: MFCD00011241
CAS Number: 7772-99-8
UN: 3260
ADR: 8 II
Merck Index: 13,08861

Order Now

Specification Test Results

Solubility 2700 g/l (20 °C)
Melting Point 246 °C
Molar Mass 189.62 g/mol
Boiling Point 623 °C (1013 hPa)
Density 3.95 g/cm3 (20 °C)
pH 2.0 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR