3,6-Dioxaoctamethylenediamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Bis-NH2-PEG2, 2,2'-(Ethylenedioxy)bis(ethylamine), 1,8-Diamino-3,6-dioxaoctane, 1,2-Bis(2-aminoethoxy)ethane

M-Clarity™ quality level = MQ200

8.18116.0025 8.18116.0100 8.18116.0025 8.18116.0100
EM8.18116.0025CS 3328.55 CAD
EM8.18116.0025 EM8.18116.0100 CA8.18116.0025 CA8.18116.0100
3,6-Dioxaoctamethylenediamine for synthesis, Sigma-Aldrich®
3,6-Dioxaoctamethylenediamine
Formula: C₆H₁₆N₂O₂
MW: 148.20 g/mol
MDL Number: MFCD00040474
CAS Number: 929-59-9
UN: 2735
ADR: 8 II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 148.20 g/mol
Boiling Point 88 °C (0.7 hPa)
Flash Point 121 °C
Density 1.01 g/cm3 (20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR