Chromium(III) chloride, anhydrous for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Chromic chloride, Chromium trichloride
M-Clarity™ quality level = MQ200
8.18090.0002
8.18090.0025
8.18090.0002
8.18090.0025
EM8.18090.0002CS
7084.72
CAD
EM8.18090.0002
EM8.18090.0025
CA8.18090.0002
CA8.18090.0025
Chromium(III) chloride, anhydrous for synthesis, Sigma-Aldrich®
Chromium (III) chloride
Formula:
Cl₃Cr MW: 158.35 g/mol Melting Pt: 1152 |
MDL Number:
MFCD00010948 CAS Number: 10025-73-7 UN: 3077 ADR: 9 III Merck Index: 12,02278 |
Specification Test Results
Solubility | (20 °C) slightly soluble |
Melting Point | 1152 °C |
Molar Mass | 158.35 g/mol |
Boiling Point | 1300 °C decomposes |
Density | 2.87 g/cm3 (25 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...