L(+)-Serine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine

M-Clarity™ quality level = MQ200

8.16020.0025 8.16020.0100 8.16020.0025 8.16020.0100
EM8.16020.0025CS 6866.68 CAD
EM8.16020.0025 EM8.16020.0100 CA8.16020.0025 CA8.16020.0100
L(+)-Serine for synthesis, Sigma-Aldrich®
L(+)-Serine
Formula: C₃H₇NO₃
MW: 105.09 g/mol
MDL Number: MFCD00064224
CAS Number: 56-45-1

Order Now

Specification Test Results

Solubility 364 g/l (20 °C)
Melting Point 215 - 225 °C decomposes
Molar Mass 105.09 g/mol
Bulk Density 570 kg/m3
pH 5 - 6 (100 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR