L(-)-Proline for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
(S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
M-Clarity™ quality level = MQ200
8.16019.0025
8.16019.0100
8.16019.0025
8.16019.0100
EM8.16019.0025CS
5732.24
CAD
EM8.16019.0025
EM8.16019.0100
CA8.16019.0025
CA8.16019.0100
L(-)-Proline for synthesis, Sigma-Aldrich®
L(-)-Proline
Formula:
C₅H₉NO₂ MW: 115.13 g/mol |
MDL Number:
MFCD00064318 CAS Number: 147-85-3 |
Specification Test Results
Solubility | 1500 g/l (20 °C) |
Melting Point | 220 - 222 °C (decomposition) |
Molar Mass | 115.13 g/mol |
Bulk Density | 500 kg/m3 |
Density | 1.35 - 1.38 g/cm3 (25 °C) |
pH | 5 - 7 (100 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...