L(+)-Lysine monohydrochloride ≥85% for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Lysine Hydrochloride, (S)-(+)-2,6-Diaminohexanoic acid monohydrochloride, L-Lysine monohydrochloride, H-Lys-OH·HCl, L-Lysine monohydrocholoride, L-Lysine Hydrochloride, (S)-(+)-Lysine monohydrochloride, (S)-(+)-2,6-Diaminocaproic acid monohydrochloride, H-Lys-OH.HCl
M-Clarity™ quality level = MQ200
8.16018.0100
8.16018.0500
8.16018.0100
8.16018.0500
EM8.16018.0100CS
7648.78
CAD
EM8.16018.0100
EM8.16018.0500
CA8.16018.0100
CA8.16018.0500
L(+)-Lysine monohydrochloride ≥85% for synthesis, Sigma-Aldrich®
L(+)-Lysine monohydrochloride
Formula:
H₂N(CH₂)₄CH(NH₂)CO₂H·HCl MW: 182.65 g/mol |
MDL Number:
MFCD00064564 CAS Number: 657-27-2 |
Specification Test Results
Solubility | 420 g/l (20 °C) |
Melting Point | 263 - 264 °C |
Molar Mass | 182.64 g/mol |
Bulk Density | 360 kg/m3 |
pH | 5.5 - 6.0 (100 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...