L(+)-Glutamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.16016.0025 8.16016.0100 8.16016.0025 8.16016.0100
EM8.16016.0025CS 4717.88 CAD
EM8.16016.0025 EM8.16016.0100 CA8.16016.0025 CA8.16016.0100
L(+)-Glutamine for synthesis, Sigma-Aldrich®
L(+)-Glutamine
Formula: C₅H₁₀N₂O₃
MDL Number: MFCD00008044
CAS Number: 56-85-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb7a812ec5d2c73a797fc464926df9eef12c5c9a&vid=15d7d87daff0fec33a974a578415e157a3278049&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 100 g/l (40 °C)
Melting Point 185 - 186 °C
Molar Mass 146.15 g/mol
Bulk Density 640 kg/m3
pH 4.0 - 6.0 (25 g/l, H2O, 20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR