L-Threonine zS, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid

MQ200

8.16012.0025 8.16012.0100 8.16012.0025 8.16012.0100
EM8.16012.0025CS 12135.98 CAD
EM8.16012.0025 EM8.16012.0100 CA8.16012.0025 CA8.16012.0100
L-Threonine zS, Sigma-Aldrich®
L-Threonine
Formula: C₄H₉NO₃
MW: 119.12 g/mol
MDL Number: MFCD00064270
CAS Number: 72-19-5

Order Now

Specification Test Results

Solubility 90 g/l (20 °C)
Melting Point 265 - 270 °C
Molar Mass 119.12 g/mol
Bulk Density 370 kg/m3
pH 5 - 6 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR