L(+)-Leucine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine

M-Clarity™ quality level = MQ200

8.16009.0025 8.16009.0100 8.16009.0025 8.16009.0100
EM8.16009.0025CS 4563.04 CAD
EM8.16009.0025 EM8.16009.0100 CA8.16009.0025 CA8.16009.0100
L(+)-Leucine for synthesis, Sigma-Aldrich®
L(+)-Leucine
Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: 300
MDL Number: MFCD00002617
CAS Number: 61-90-5
Merck Index: 14,05451

Order Now

Specification Test Results

Solubility 24 g/l (20 °C)
Melting Point 300 °C (decomposition)
Molar Mass 131.18 g/mol
Bulk Density 300 - 400 kg/m3
Density 1.29 g/cm3 (20 °C)
pH 5.5 - 6.5 (20 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR