L(+)-Isoleucine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: L-Isoleucine, (2S,3S)-(+)-Isoleucine, H-Ile-OH

M-Clarity™ quality level = MQ200

8.16008.0010 8.16008.0025 8.16008.0010 8.16008.0025
EM8.16008.0010CS 8506.72 CAD
EM8.16008.0010 EM8.16008.0025 CA8.16008.0010 CA8.16008.0025
L(+)-Isoleucine for synthesis, Sigma-Aldrich®
L(+)-Isoleucine
Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
MDL Number: MFCD00064222
CAS Number: 73-32-5

Order Now

Specification Test Results

Solubility 32.1 g/l (20 °C)
Melting Point 279 - 280 °C (decomposition)
Molar Mass 131.17 g/mol
Bulk Density 280 kg/m3
pH 6.0 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR