L(+)-Histidine monohydrochloride monohydrate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
L-Histidine monohydrochloride monohydrate, (2S)-(+)-Histidine monohydrochloride monohydrate, L-Histidine hydrochloride monohydrate, (S)-(+)-Histidine monohydrochloride monohydrate, His HCl monohydrate, H-His-OH.HCl.H2O, H-His-OH·HCl·H₂O, L-3-Imidazol-4-ylalanine monohydrochloride monohydrate, L-Histidine hydrochloride hydrate
M-Clarity™ quality level = MQ200
8.16005.0025
8.16005.0100
8.16005.0025
8.16005.0100
EM8.16005.0025CS
6174.64
CAD
EM8.16005.0025
EM8.16005.0100
CA8.16005.0025
CA8.16005.0100
L(+)-Histidine monohydrochloride monohydrate for synthesis, Sigma-Aldrich®
L(+)-Histidine monohydrochloride monohydrate
Formula:
C₆H₉N₃O₂·HCl·H₂O MW: 209.63 g/mol |
MDL Number:
MFCD00151027 CAS Number: 5934-29-2 |
Specification Test Results
Solubility | 169.9 g/l (20 °C) |
Melting Point | 259 °C (decomposition) |
Molar Mass | 209.63 g/mol |
Bulk Density | 770 kg/m3 |
pH | 3.5 - 4.5 (100 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...