L(+)-Aspartic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
(S)-(+)-Aminosuccinic acid (S)-Aminobutanedioic acid (S)-Aminosuccinic acid H-Asp-OH L-Asparaginic acid L(+)-Aspartic acid, (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, L-Aspartic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D
M-Clarity™ quality level = MQ200
8.16003.0100
8.16003.0500
8.16003.0100
8.16003.0500
EM8.16003.0100CS
6624.94
CAD
EM8.16003.0100
EM8.16003.0500
CA8.16003.0100
CA8.16003.0500
L(+)-Aspartic acid for synthesis, Sigma-Aldrich®
L(+)-Aspartic acid
Formula:
C₄H₇NO₄ MW: 133.1 g/mol |
MDL Number:
MFCD00002616 CAS Number: 56-84-8 |
Specification Test Results
Assay (acidimetric) | ≥ 99.0 % |
Identity (IR) | passes test |
Spec. rotation α²0/D (c=8 in HCl 6 mol/l) | +24.5 - +26.0 ° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...