Aluminium, grit for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Aluminum

M-Clarity™ quality level = MQ200

8.14917.0100 8.14917.0100 8.14917.1000 8.14917.1000
CA8.14917.0100CS 10560.62 CAD
CA8.14917.0100 EM8.14917.0100 EM8.14917.1000 CA8.14917.1000
Aluminium, grit for synthesis, Sigma-Aldrich®
Aluminium
Formula: Al
MW: 26.98 g/mol
MDL Number: MFCD00134029
CAS Number: 7429-90-5
UN: 1396
ADR: 4.3,II

Order Now

Specification Test Results

Solubility (20 °C) (reaction)
Molar Mass 26.98 g/mol
Hazard Class 4.3
Hazard Class Description Dangerous When Wet Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR