DL-Malic acid zS, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: DL-Hydroxysuccinic acid, (±)-Malic acid, (±)-Hydroxy-1,4-butanedioic acid, (±)-Hydroxysuccinic acid, Hydroxybutanedioic acid, 2-Hydroxysuccinic acid

MQ200

8.14737.1000 8.14737.5000 8.14737.1000 8.14737.5000
EM8.14737.1000CS 854.78 CAD
EM8.14737.1000 EM8.14737.5000 CA8.14737.1000 CA8.14737.5000
DL-Malic acid zS, Sigma-Aldrich®
DL-Malic acid
Formula: HOOCCH(OH)CH₂COOH
MW: 134.09 g/mol
Boiling Pt: 306
MDL Number: MFCD00064212
CAS Number: 617-48-1
Merck Index: 13,05730
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=920b54632a869bc5df22624fb9fecd4d11cae65e&vid=278193678126274041defdc0bd71c7c40efb3085&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (acidimetric) ≥ 99.0 %
Identity (IR) passes test
Melting range (lower value) ≥ 128 °C
Melting range (upper value) ≤ 132 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR