Magnesium chloride, anhydrous for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.14733.0500 8.14733.0100 8.14733.0500 8.14733.0100
CA8.14733.0500EA 181 CAD
CA8.14733.0500 EM8.14733.0100 EM8.14733.0500 CA8.14733.0100
Magnesium chloride, anhydrous for synthesis, Sigma-Aldrich®
Magnesium chloride
Formula: MgCl₂
MDL Number: MFCD00011106
CAS Number: 7786-30-3

Order Now

Specification Test Results

Solubility 542 g/l (20 °C)
Melting Point 712 °C
Molar Mass 95.22 g/mol
Boiling Point 1412 °C (1013 hPa)
Density 2.32 g/cm3 (20 °C)
pH =7 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR