Brij® 58 stabilised for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Polyoxyethylene (20) cetyl ether, Polyethylene glycol hexadecyl ether

Stabilization: Stabilised with BHA (2-tert-butyl-4-methoxyphenol ) and citric acid
M-Clarity™ quality level = MQ200

8.14727.0250 8.14727.1000 8.14727.0250 8.14727.1000
EM8.14727.0250CS 587.76 CAD
EM8.14727.0250 EM8.14727.1000 CA8.14727.0250 CA8.14727.1000
Brij® 58 stabilised for synthesis, Sigma-Aldrich®
Brij® 58
Formula: C₅₆H₁₁₄O₂₁
MW: 1123.51 g/mol
MDL Number: MFCD00080892
CAS Number: 9004-95-9

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 38 - 42 °C
Flash Point 149 °C
Density 1 g/cm3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR