2-(2-Ethoxyethoxy)ethyl acetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Diethylene glycol monoethyl ether acetate

M-Clarity™ quality level = MQ200

8.14635.0250 8.14635.1000 8.14635.0250 8.14635.1000
EM8.14635.0250CS 1153.4 CAD
EM8.14635.0250 EM8.14635.1000 CA8.14635.0250 CA8.14635.1000
2-(2-Ethoxyethoxy)ethyl acetate for synthesis, Sigma-Aldrich®
2-(2-Ethoxyethoxy)ethyl acetate
Formula: C₈H₁₆O₄
MW: 176.21 g/mol
Boiling Pt: 210
Melting Pt: -25
MDL Number: MFCD00041928
CAS Number: 112-15-2

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -25 °C
Molar Mass 176.21 g/mol
Boiling Point 217 - 218 °C
Vapor Pressure 0.3 hPa (20 °C)
Flash Point 100 °C
Explosion Limit 1.5 %(V)
Density 1.01 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 310 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR