Triethylene glycol monomethyl ether for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
mPEG3-Alcohol, 2-(2-(2-Methoxyethoxy)ethoxy)ethanol, Methyl triglycol, m-PEG3-OH
M-Clarity™ quality level = MQ200
8.14587.0250
8.14587.1000
8.14587.0250
8.14587.1000
EM8.14587.0250CS
742.6
CAD
EM8.14587.0250
EM8.14587.1000
CA8.14587.0250
CA8.14587.1000
Triethylene glycol monomethyl ether for synthesis, Sigma-Aldrich®
Triethylene glycol monomethyl ether
Formula:
C₇H₁₆O₄ MW: 164.2 g/mol |
MDL Number:
MFCD00002875 CAS Number: 112-35-6 |
Specification Test Results
Solubility | (20 °C) soluble |
Molar Mass | 164.2 g/mol |
Boiling Point | 245 - 255 °C (1013 hPa) |
Vapor Pressure |
|
Flash Point | 133 °C |
Density | 1.04 g/cm3 (20 °C) |
pH | 5 - 7 (H2O, 20 °C) |
Ignition Point | 210 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...