(±)-Citronellal for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.14575.0100 8.14575.0500 8.14575.0100 8.14575.0500
EM8.14575.0100CS 1273.48 CAD
EM8.14575.0100 EM8.14575.0500 CA8.14575.0100 CA8.14575.0500
(±)-Citronellal for synthesis, Sigma-Aldrich®
(±)-Citronellal
Formula: C₁₀H₁₈O
MDL Number: MFCD00038090
CAS Number: 106-23-0
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Molar Mass 154.25 g/mol
Boiling Point 208 °C (1013 hPa)
Vapor Pressure 14 hPa (88 °C)
Flash Point 77 °C
Refractive Index 1.446 (20 °C)
Explosion Limit 1.2 - 4.5 %(V)
Density 0.85 g/cm3 (20 °C)
pH 7 (H2O)
Ignition Point 202 °C
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR