2-Chloroethyldiisopropylammonium chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.14553.0100 8.14553.0250 8.14553.0100 8.14553.0250
EM8.14553.0100CS 3062.04 CAD
EM8.14553.0100 EM8.14553.0250 CA8.14553.0100 CA8.14553.0250
2-Chloroethyldiisopropylammonium chloride for synthesis, Sigma-Aldrich®
2-Chloroethyldiisopropylammonium chloride
Formula: C₈H₁₉Cl₂N
MDL Number: MFCD00012496
CAS Number: 4261-68-1
UN: 2811
ADR: 6.1,I

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 130 - 133 °C
Molar Mass 200.15 g/mol
pH 4.0 - 4.5 (10 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR