Cetrimonium bromide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide

M-Clarity™ quality level = MQ200

8.14119.0100 8.14119.0500 8.14119.0100 8.14119.0500
EM8.14119.0100CS 802.64 CAD
EM8.14119.0100 EM8.14119.0500 CA8.14119.0100 CA8.14119.0500
Cetrimonium bromide for synthesis, Sigma-Aldrich®
Cetrimonium bromide
Formula: CH₃(CH₂)₁₅N(Br)(CH₃)₃
MW: 364.45 g/mol
Flash Pt: 244
MDL Number: MFCD00011772
CAS Number: 57-09-0
UN: 3077
ADR: 9 III
Merck Index: 13,02034

Order Now

Specification Test Results

Solubility 0.192 g/l (20 °C)
Melting Point 250 - 256 °C
Molar Mass 364.45 g/mol
Bulk Density 390 kg/m3
pH 5 - 7 (50 g/l, H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR