Glyoxylic acid 50% in water for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.14084.0250 8.14084.1000 8.14084.0250 8.14084.1000
EM8.14084.0250CS 1202.38 CAD
EM8.14084.0250 EM8.14084.1000 CA8.14084.0250 CA8.14084.1000
Glyoxylic acid 50% in water for synthesis, Sigma-Aldrich®
Glyoxylic acid
Formula: C₂H₂O₃
MDL Number: MFCD00006958
CAS Number: 298-12-4
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Boiling Point 111 °C (1013 hPa)
Vapor Pressure 10 hPa (25 °C)
Flash Point 115 °C
Refractive Index 1.4149 (20 °C, 589 nm)
Density 1.34 g/cm3 (20 °C)
pH 0.3 (H2O, 19.5 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR