Dibenzo-24-crown-8-(dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosane) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.13755.0001 8.13755.0005 8.13755.0001 8.13755.0005
EM8.13755.0001CS 231296.2 CAD
EM8.13755.0001 EM8.13755.0005 CA8.13755.0001 CA8.13755.0005
Dibenzo-24-crown-8-(dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosane) for synthesis, Sigma-Aldrich®
Dibenzo-24-crown-8 (Dibenzo-1,4,7,10,13,16,19,22-octaoxacyclotetracosane)
Formula: C₂₄H₃₂O₈
MDL Number: MFCD00005101
CAS Number: 14174-09-5

Order Now

Specification Test Results

Melting Point 100 - 103 °C
Molar Mass 448.52 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR