Ethyl acetoacetate ≥98.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Acetoacetic acid ethyl ester, EAA

M-Clarity™ quality level = MQ200

8.09622.0100 8.09622.2500 8.09622.0100 8.09622.1000 8.09622.2500 8.09622.9025
EM8.09622.0100CS 3009.9 CAD
EM8.09622.0100 EM8.09622.2500 CA8.09622.0100 CA8.09622.1000 CA8.09622.2500 CA8.09622.9025
Ethyl acetoacetate ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Ethyl acetoacetate
Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Melting Pt: –53.3 °C
Density: 1.03 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009199
CAS Number: 141-97-9

Order Now

Specification Test Results

Solubility 116 g/l (20 °C)
Melting Point -40 °C
Molar Mass 130.14 g/mol
Boiling Point 180.0 °C (1013 hPa)
Vapor Pressure 1 hPa (20 °C)
Flash Point 65 °C
Refractive Index 1.419 (20 °C, 589 nm)
Explosion Limit 1.0 - 54 %(V)
Density 1.03 g/cm3 (20 °C)
pH 4.0 (110 g/l, H2O, 20 °C)
Ignition Point 280 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR