1,2,4-Triazole for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 1,2,4-1H-Triazole, 1H-1,2,4-Triazole

M-Clarity™ quality level = MQ200

8.08388.0025 8.08388.0100 8.08388.0025 8.08388.0100
EM8.08388.0025CS 6040.34 CAD
EM8.08388.0025 EM8.08388.0100 CA8.08388.0025 CA8.08388.0100
1,2,4-Triazole for synthesis, Sigma-Aldrich®
1,2,4-Triazole
Formula: C₂H₃N₃
MW: 69.07 g/mol
MDL Number: MFCD00005228
CAS Number: 288-88-0

Order Now

Specification Test Results

Solubility 547 g/l (22 °C)
Melting Point 119 - 122 °C
Molar Mass 69.07 g/mol
Bulk Density 550 kg/m3
Boiling Point 259 - 261 °C (1013 hPa)
Density 1.15 g/cm3 (130 °C)
pH 8 (10 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR