Trityl chloride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Trt-Cl, Chlorotriphenylmethane, Triphenylmethyl chloride, (Chloromethanetriyl)tribenzene, Triphenylchloromethane
M-Clarity™ quality level = MQ200
8.08348.0250
8.08348.1000
8.08348.0250
8.08348.1000
EM8.08348.0250CS
2739.72
CAD
EM8.08348.0250
EM8.08348.1000
CA8.08348.0250
CA8.08348.1000
Trityl chloride for synthesis, Sigma-Aldrich®
Trityl chloride
Formula:
C₁₉H₁₅Cl MW: 278.78 g/mol Boiling Pt: 374 |
MDL Number:
MFCD00000813 CAS Number: 76-83-5 UN: 1759 ADR: 8 II |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Melting Point | 109 - 112 °C |
Molar Mass | 278.78 g/mol |
Boiling Point | 230 - 235 °C (27 hPa) |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...