Ethyl toluene-4-sulfonate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Ethyl 4-toluenesulphonate, p-Toluenesulfonic acid ethyl ester, Ethyl 4-methylbenzenesulfonate, Toluene-4-sulfonic acid ethyl ester, Ethyl p-toluenesulphonate, Ethyl p-toluenesulfonate, p-Toluenesulphonic acid ethyl ester, Ethyl 4-toluenesulfonate, Ethyl 4-methylbenzenesulphonate, Toluene-4-sulphonic acid ethyl ester
M-Clarity™ quality level = MQ200
8.08332.0100
8.08332.0500
8.08332.5000
8.08332.0100
8.08332.0500
8.08332.5000
EM8.08332.0100CS
1932.34
CAD
EM8.08332.0100
EM8.08332.0500
EM8.08332.5000
CA8.08332.0100
CA8.08332.0500
CA8.08332.5000
Ethyl toluene-4-sulfonate for synthesis, Sigma-Aldrich®
Ethyl toluene-4-sulphonate
Formula:
C₉H₁₂O₃S MW: 200.26 g/mol Boiling Pt: 158 |
MDL Number:
MFCD00009100 CAS Number: 80-40-0 UN: 2811 Merck Index: 13,03889 |
Specification Test Results
Solubility | 1.24 g/l (25 °C) |
Melting Point | 30 - 32 °C |
Molar Mass | 200.25 g/mol |
Boiling Point | 221 °C (1013 hPa) |
Vapor Pressure | 0.6 hPa (14 °C) |
Flash Point | 158 °C |
Density | 1.174 g/cm3 (20 °C) |
Ignition Point | 500 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...