Titanium(III) chloride ∼30% in hydrochloric acid 10% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Titanium trichloride

M-Clarity™ quality level = MQ200

8.08308.0100 8.08308.0500 8.08308.0100 8.08308.0500
EM8.08308.0100CS 1952.88 CAD
EM8.08308.0100 EM8.08308.0500 CA8.08308.0100 CA8.08308.0500
Titanium(III) chloride ∼30% in hydrochloric acid 10% for synthesis, Sigma-Aldrich®
Titanium (III) chloride
Formula: Cl₃Ti
MW: 154.24
Boiling Pt: 960
Melting Pt: 440
Density: 1.32
MDL Number: MFCD00011266
CAS Number: 7705-07-9
EINECS: 231-728-9
UN: 1789
ADR: 8 II
Merck Index: 12,09620

Order Now

Specification Test Results

Density 1.017 g/cm3 (20 °C)
pH 0.5 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR