Titanium(III) chloride ∼15% in hydrochloric acid 10% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Titanium trichloride

M-Clarity™ quality level = MQ200

8.08307.1000 8.08307.0100 8.08307.0500 8.08307.1000 8.08307.2500 8.08307.0500 8.08307.2500
CA8.08307.1000EA 329 CAD
CA8.08307.1000 CA8.08307.0100 EM8.08307.0500 EM8.08307.1000 EM8.08307.2500 CA8.08307.0500 CA8.08307.2500
Titanium(III) chloride ∼15% in hydrochloric acid 10% for synthesis, Sigma-Aldrich®
Titanium (III) chloride
Formula: Cl₃Ti
MW: 154.24
Boiling Pt: 960
Melting Pt: 440
Density: 1.21 g/cm³ (20 °C)
MDL Number: MFCD00011266
CAS Number: 7705-07-9
EINECS: 231-728-9
UN: 1789
ADR: 8 II
Merck Index: 12,09620

Order Now

Specification Test Results

Assay (cerimetric TiCl₃) 14.5 - 15.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR