Thiophene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.08157.0005 8.08157.0250 8.08157.1000 8.08157.0005 8.08157.0250 8.08157.1000
EM8.08157.0005CS 3389.1 CAD
EM8.08157.0005 EM8.08157.0250 EM8.08157.1000 CA8.08157.0005 CA8.08157.0250 CA8.08157.1000
Thiophene for synthesis, Sigma-Aldrich®
Thiophene
Formula: C₄H₄S
Boiling Pt: 84.4 °C (1013 hPa)
Melting Pt: –38 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: –9 °C
Storage Temperature: Ambient
MDL Number: MFCD00005413
CAS Number: 110-02-1
EINECS: 203-729-4
UN: 2414
ADR: 3,II
Merck Index: 12,09490

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -38 °C
Molar Mass 84.14 g/mol
Boiling Point 83 - 84 °C (1013 mbar)
Vapor Pressure 80 hPa (20 °C)
Flash Point -9 °C
Refractive Index 1.5289 (20 °C, 589 nm)
Explosion Limit 1.5 - 12.5 %(V)
Density 1.06 g/cm3 (20 °C)
Ignition Point 395 °C
Saturation concentration 276 g/m3 (20 °C) Air
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR