3-Pentanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.07502.0100 8.07502.0500 8.07502.0100 8.07502.0500
EM8.07502.0100CS 1756.96 CAD
EM8.07502.0100 EM8.07502.0500 CA8.07502.0100 CA8.07502.0500
3-Pentanol for synthesis, Sigma-Aldrich®
3-Pentanol
Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 116
Melting Pt: –8 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 33
Storage Temperature: Ambient
MDL Number: MFCD00004574
CAS Number: 584-02-1
EINECS: 209-526-7
UN: 1105
ADR: 3 III
Merck Index: 13,07197
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ec7192926b50615dfac930445c63174c6221a44&vid=a2f1fa3f1ffa1ecbfb6a5dd1d0f29da27d1d8737&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 55 g/l (30 °C)
Melting Point -8 °C
Molar Mass 88.14 g/mol
Boiling Point 115 - 116 °C (1013 hPa)
Vapor Pressure 11.69 hPa (25 °C)
Flash Point 33 °C
Refractive Index 1.4096 (20 °C, 589 nm)
Explosion Limit 1.2 - 8 %(V)
Density 0.82 g/cm3 (20 °C)
Ignition Point 360 °C
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR