(±)-2-Pentanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.07501.0100 8.07501.0500 8.07501.0100 8.07501.0500
EM8.07501.0100CS 2576.98 CAD
EM8.07501.0100 EM8.07501.0500 CA8.07501.0100 CA8.07501.0500
(±)-2-Pentanol for synthesis, Sigma-Aldrich®
(±)-2-Pentanol
Formula: C₅H₁₂O
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00004579
CAS Number: 6032-29-7
EINECS: 227-907-6
UN: 1105
ADR: 3,III
Merck Index: 12,07258
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc37771666c375a7768be6819fbc64e80617a202&vid=cc73accd09f3b4e43eedad1fe603cf52c119ff25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 166 g/l (20 °C)
Melting Point -50 °C
Molar Mass 88.14 g/mol
Boiling Point 119 °C (1013 hPa)
Vapor Pressure 3 hPa (20 °C)
Flash Point 41 °C
Refractive Index 1.4055 (20 °C, 589 nm)
Explosion Limit 1.2 - 10.5 %(V)
Density 0.81 g/cm3 (20 °C)
Ignition Point 330 °C
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR