Polyethylene glycol 4,000 for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.07490.1000 8.07490.5000 8.07490.1000 8.07490.5000
EM8.07490.1000CS 655.7 CAD
EM8.07490.1000 EM8.07490.5000 CA8.07490.1000 CA8.07490.5000
Polyethylene glycol 4,000 for synthesis, Sigma-Aldrich®
Polyethylene glycol 4,000
Formula: H(OCH₂CH₂)nOH
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 53…58 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD01779612
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Specification Test Results

Solubility 500 g/l (20 °C)
Melting Point 53 - 58 °C
Molar Mass 3500 - 4500 g/mol
Bulk Density 400 - 500 kg/m3
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 250 °C
Density 1.2 g/cm3 (20 °C)
pH 5 - 7 (100 g/l, H2O, 20 °C)
Ignition Point 320 °C DIN 51794
Viscosity kinematic 76 - 110 mm2/s (98 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR