Polyethylene glycol 1,500 for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: PEG 1500, PEG 1,500

M-Clarity™ quality level = MQ200

8.07489.1000 8.07489.5000 8.07489.1000 8.07489.5000
EM8.07489.1000CS 741.02 CAD
EM8.07489.1000 EM8.07489.5000 CA8.07489.1000 CA8.07489.5000
Polyethylene glycol 1,500 for synthesis, Sigma-Aldrich®
Polyethylene glycol 1,500
Formula: H(OCH₂CH₂)nOH
MW: 1500 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 44…48 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 260
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Specification Test Results

Solubility 500 g/l (20 °C)
Melting Point 44 - 48 °C
Molar Mass 8000 g/mol
Bulk Density 400 - 500 kg/m3
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 260 °C
Density 1.2 g/cm3 (20 °C)
pH 4 - 7 (100 g/l, H2O, 20 °C)
Ignition Point 420 °C
Viscosity kinematic 33 - 39 mm2/s (20 °C) (50% solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR