Polyethylene glycol 1,000 for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.07488.1000
8.07488.5000
8.07488.1000
8.07488.5000
EM8.07488.1000CS
722.06
CAD
EM8.07488.1000
EM8.07488.5000
CA8.07488.1000
CA8.07488.5000
Polyethylene glycol 1,000 for synthesis, Sigma-Aldrich®
Polyethylene glycol 1,000
Formula:
H(OCH₂CH₂)nOH Boiling Pt: ∼250 °C (1013 hPa) Melting Pt: 33…40 °C Density: 1.2 g/cm³ (20 °C) Flash Pt: 260 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 500-038-2 Merck Index: 13,07651 |
Specification Test Results
Solubility | 750 g/l (20 °C) |
Melting Point | 33 - 40 °C |
Vapor Pressure |
|
Flash Point | 260 °C |
Density | 1.2 g/cm3 (20 °C) |
pH | 4 - 7 (100 g/l, H2O, 20 °C) |
Ignition Point | 420 °C |
Viscosity kinematic | 22 - 27 mm2/s (20 °C) (50% solution) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...