2-Phenoxyethanol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.07291.1000
8.07291.9025
8.07291.1000
8.07291.2500
8.07291.5000
8.07291.2500
8.07291.5000
CA8.07291.1000EA
63
CAD
CA8.07291.1000
CA8.07291.9025
EM8.07291.1000
EM8.07291.2500
EM8.07291.5000
CA8.07291.2500
CA8.07291.5000
2-Phenoxyethanol for synthesis, Sigma-Aldrich®
2-Phenoxyethanol
Formula:
C₆H₅OCH₂CH₂OH Boiling Pt: 245 °C (1013 hPa) Melting Pt: 13 °C Density: 1.1094 g/cm³ (20 °C) Flash Pt: 121 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00002857 CAS Number: 122-99-6 EINECS: 204-589-7 Merck Index: 12,07410 |
|
Specification Test Results
Solubility | 24 g/l (20 °C) |
Melting Point | 13 °C |
Molar Mass | 138.16 g/mol |
Boiling Point | 244 - 250 °C (1013 hPa) |
Vapor Pressure | 0.04 hPa (20 °C) |
Flash Point | 121 °C DIN 51758 |
Refractive Index | 1.534 (20 °C, 589 nm) |
Explosion Limit | 1.4 - 9.0 %(V) |
Density | 1.11 g/cm3 (20 °C) |
pH | 7 (10 g/l, H2O, 23 °C) |
Ignition Point | 535 °C DIN 51794 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...