Palladium(II) chloride Pd 59%, anhydrous for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.07110.0001 8.07110.0005 8.07110.0010 8.07110.0001 8.07110.0005 8.07110.0010 8.07110.8500
EM8.07110.0001CS 64960.12 CAD
EM8.07110.0001 EM8.07110.0005 EM8.07110.0010 CA8.07110.0001 CA8.07110.0005 CA8.07110.0010 CA8.07110.8500
Palladium(II) chloride Pd 59%, anhydrous for synthesis, Sigma-Aldrich®
Palladium (II) chloride
Formula: PdCl₂
Melting Pt: 678 °C
Density: 4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003558
CAS Number: 7647-10-1
EINECS: 231-596-2
UN: 3260
ADR: 8,II
Merck Index: 13,07058

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 680 °C
Molar Mass 177.33 g/mol
Density 4.0 g/cm3 (20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR