Propiononitrile for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.07068.0100
8.07068.0500
8.07068.0100
8.07068.0500
8.07068.6190
EM8.07068.0100CS
1420.42
CAD
EM8.07068.0100
EM8.07068.0500
CA8.07068.0100
CA8.07068.0500
CA8.07068.6190
Propiononitrile for synthesis, Sigma-Aldrich®
Propiononitrile
Formula:
C₃H₅N Boiling Pt: 97.2 °C (1013 hPa) Melting Pt: –93 °C Density: 0.782 g/cm³ (20 °C) Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001948 CAS Number: 107-12-0 EINECS: 203-464-4 UN: 2404 ADR: 3 (6.1),II Merck Index: 12,08012 |
Specification Test Results
Solubility | 99.4 g/l (20 °C) |
Melting Point | -92 °C |
Molar Mass | 55.08 g/mol |
Boiling Point | 97 °C (1013 hPa) |
Vapor Pressure | 50 hPa (20 °C) |
Flash Point | 12 °C |
Refractive Index | 1.366 (20 °C, 589 nm) |
Explosion Limit | 3.1 - 14 %(V) |
Density | 0.782 g/cm3 (20 °C) |
pH | 10 (50 g/l, H2O, 20 °C) |
Ignition Point | 515 °C |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...