Caprylaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Octanal, Octylaldehyde, Octaldehyde

M-Clarity™ quality level = MQ200

8.06901.0100 8.06901.0500 8.06901.0100 8.06901.0500
EM8.06901.0100CS 1058.6 CAD
EM8.06901.0100 EM8.06901.0500 CA8.06901.0100 CA8.06901.0500
Caprylaldehyde for synthesis, Sigma-Aldrich®
Caprylaldehyde
Formula: C₈H₁₆O
Boiling Pt: ∼171 °C (1013 hPa)
Melting Pt: ∼12 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00007029
CAS Number: 124-13-0
EINECS: 204-683-8
UN: 1989
ADR: 3,III
Merck Index: 13,01772

Order Now

Specification Test Results

Solubility 0.21 g/l (20 °C)
Melting Point 12 - 15 °C
Molar Mass 128.21 g/mol
Boiling Point 167 - 169 °C (1013 hPa)
Vapor Pressure 1 hPa (20 °C)
Flash Point 56 °C
Refractive Index 1.419 (20 °C, 589 nm)
Explosion Limit 1.0 - 6.5 %(V)
Density 0.82 g/cm3 (20 °C)
Ignition Point 190 °C
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR