Ethyl pyruvate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.06617.0100
8.06617.0500
8.06617.0100
8.06617.0500
EM8.06617.0100CS
1463.08
CAD
EM8.06617.0100
EM8.06617.0500
CA8.06617.0100
CA8.06617.0500
Ethyl pyruvate for synthesis, Sigma-Aldrich®
Ethyl pyruvate
Formula:
C₅H₈O₃ Boiling Pt: 144…147 °C (1013 hPa) Melting Pt: –50 °C Density: 1.05 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009123 CAS Number: 617-35-6 EINECS: 210-511-2 UN: 3272 ADR: 3,III |
Specification Test Results
Solubility | 10 g/l (20 °C) |
Melting Point | -58 °C |
Molar Mass | 116.11 g/mol |
Boiling Point | 144 - 146 °C (1013 hPa) |
Flash Point | 52 °C |
Refractive Index | 1.4056 (20 °C, 589 nm) |
Density | 1.05 g/cm3 (20 °C) |
pH | 2.2 (300 g/l, H2O, 20 °C) |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...