Morpholine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.06127.0100
8.06127.0500
8.06127.1000
8.06127.2500
8.06127.0100
8.06127.0500
8.06127.1000
8.06127.2500
EM8.06127.0100CS
2507.46
CAD
EM8.06127.0100
EM8.06127.0500
EM8.06127.1000
EM8.06127.2500
CA8.06127.0100
CA8.06127.0500
CA8.06127.1000
CA8.06127.2500
Morpholine for synthesis, Sigma-Aldrich®
Morpholine
Formula:
C₄H₉NO Boiling Pt: 128.9 °C (1013 hPa) Melting Pt: –5 °C Density: 1 g/cm³ (20 °C) Flash Pt: 38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005972 CAS Number: 110-91-8 EINECS: 203-815-1 UN: 2054 ADR: 8 (3),I Merck Index: 12,06362 |
|
Specification Test Results
Solubility | (20 °C) soluble |
Molar Mass | 87.12 g/mol |
Boiling Point | 128 - 130 °C (1013 hPa) |
Vapor Pressure | 10 hPa (20 °C) |
Flash Point | 32 °C |
Density | 1.00 g/cm3 (20 °C) |
pH | 10.6 (5 g/l, H2O, 20 °C) |
Ignition Point | 275 °C |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...