3-Methoxypropiononitrile for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 3-Methoxypropanenitrile, 3-Methoxypropionitrile

M-Clarity™ quality level = MQ200

8.05891.0100 8.05891.0500 8.05891.0100 8.05891.0500
EM8.05891.0100CS 1036.48 CAD
EM8.05891.0100 EM8.05891.0500 CA8.05891.0100 CA8.05891.0500
3-Methoxypropiononitrile for synthesis, Sigma-Aldrich®
3-Methoxypropiononitrile
Formula: C₄H₇NO
MW: 85.11 g/mol
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: –63 °C
Density: 0.94 g/cm³ (20 °C)
MDL Number: MFCD00001958
CAS Number: 110-67-8
EINECS: 203-790-7

Order Now

Specification Test Results

Solubility 335 g/l (20 °C)
Melting Point -63 °C
Molar Mass 85.11 g/mol
Boiling Point 164 - 165 °C (1013 hPa)
Vapor Pressure 9 hPa (50 °C)
Flash Point 66 °C
Refractive Index 1.4035 (20 °C, 589 nm)
Explosion Limit 1.9 - 15.3 %(V)
Density 0.94 g/cm3 (20 °C)
pH 6.8 (111 g/l, H2O, 25 °C)
Ignition Point 390 °C DIN 51794

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR