2,2'-Methyliminodiethanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.05851.0100 8.05851.1000 8.05851.0100 8.05851.1000
EM8.05851.0100CS 1230.82 CAD
EM8.05851.0100 EM8.05851.1000 CA8.05851.0100 CA8.05851.1000
2,2'-Methyliminodiethanol for synthesis, Sigma-Aldrich®
2,2'-Methyliminodiethanol
Formula: H₃CN(CH₂CH₂OH)₂
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: –21 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 137 °C
MDL Number: MFCD00002848
CAS Number: 105-59-9
EINECS: 203-312-7

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -21 °C
Molar Mass 119.16 g/mol
Boiling Point 243 °C (1013 hPa)
Vapor Pressure 0.026 hPa (40 °C)
Flash Point 137 °C
Refractive Index 1.4685 (20 °C, 589 nm)
Explosion Limit 0.9 - 8.4 %(V)
Density 1.04 g/cm3 (20 °C)
pH 11.5 (100 g/l, H2O, 20 °C)
Ignition Point 265 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR