Hydrazinium hydroxide ∼ 100% N₂H₅OH for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.04608.0250
8.04608.1000
8.04608.0250
8.04608.1000
EM8.04608.0250CS
4291.28
CAD
EM8.04608.0250
EM8.04608.1000
CA8.04608.0250
CA8.04608.1000
Hydrazinium hydroxide ∼ 100% N₂H₅OH for synthesis, Sigma-Aldrich®
Hydrazinium hydroxide
Formula:
NH₂NH₂·H₂O Boiling Pt: 117.2 °C (1013 hPa) Melting Pt: –51.7 °C Density: 1.03 g/cm³ (20 °C) Flash Pt: 75 °C |
MDL Number:
MFCD00149931 CAS Number: 7803-57-8 EINECS: 206-114-9 UN: 2030 ADR: 8 (6.1),II Merck Index: 13,04790 |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | -51.7 °C |
Molar Mass | 50.06 g/mol |
Boiling Point | 120.5 °C (1013 hPa) |
Vapor Pressure | 20 hPa (20 °C) |
Flash Point | 33 °C |
Explosion Limit |
|
Density | 1.03 g/cm3 (20 °C) |
pH |
|
Ignition Point | 280 °C |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...