4-Hydroxy-4-methyl-2-pentanone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Diacetone alcohol

M-Clarity™ quality level = MQ200

8.04512.0100 8.04512.1000 8.04512.2500 8.04512.0100 8.04512.2500 8.04512.6025
EM8.04512.0100CS 1346.16 CAD
EM8.04512.0100 EM8.04512.1000 EM8.04512.2500 CA8.04512.0100 CA8.04512.2500 CA8.04512.6025
4-Hydroxy-4-methyl-2-pentanone for synthesis, Sigma-Aldrich®
4-Hydroxy-4-methyl-2-pentanone
Formula: (CH₃)₂C(OH)CH₂COCH₃
MW: 116.16 g/mol
Boiling Pt: 166
Melting Pt: -44
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 58
Storage Temperature: Ambient
MDL Number: MFCD00004471
CAS Number: 123-42-2
EINECS: 204-626-7
UN: 1148
ADR: 3 III
REACH: 01-2119473975-21
Merck Index: 13,02983
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2da78bff918d252787ee81ab8857bd3b19e51ac&vid=d8a0cccff2cac15b826ba733a013451ac11e3b51&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -47 °C
Molar Mass 116.16 g/mol
Boiling Point 168 °C
Vapor Pressure 1.1 hPa (20 °C)
Flash Point 58 °C
Refractive Index 1.4233 (20 °C, 589 nm)
Explosion Limit 1.8 - 6.9 %(V)
Density 0.94 g/cm3 (20 °C)
Ignition Point 640 °C
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR